Skip to Content
Merck
All Photos(2)

Key Documents

74593

Supelco

Obidoxime chloride

analytical standard

Synonym(s):

1,1′-(Oxydimethylene)bis(pyridinium-4-carbaldoxime) dichloride, Bis(4-formylpyridiniomethyl) ether dioxime, Toxogonin

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$215.00

$215.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$215.00

About This Item

Empirical Formula (Hill Notation):
C14H16Cl2N4O3
CAS Number:
Molecular Weight:
359.21
Beilstein:
4117377
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$215.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water (calculated from elemental analysis)

format

neat

SMILES string

[Cl-].[Cl-].O\N=C\c1cc[n+](COC[n+]2ccc(cc2)\C=N\O)cc1

InChI

1S/C14H14N4O3.2ClH/c19-15-9-13-1-5-17(6-2-13)11-21-12-18-7-3-14(4-8-18)10-16-20;;/h1-10H,11-12H2;2*1H

InChI key

ZIFJVJZWVSPZLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Obidoxime chloride is a drug, belonging to the group of oximes.[1] It is found to be useful for the treatment of organophosphorus (OP) poisoning, since it has the ability to reverse the binding of organophosphorus compounds to the enzyme acetylcholinesterase (AChE).[2][3]

Application

Obidoxime chloride may be used as a reference standard for the determination of obidoxime in a parenteral injection device using high-performance liquid chromatography (HPLC).[4]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Antidote for organophosphate nerve agent poisoning[5]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple high-performance liquid chromatographic method for determination of atropine and obidoxime in a parenteral injection device.
Journal of Chromatography A, 1057(1-2), 237-239 (2004)
Best Synthetic Methods: Organophosphorus (V) Chemistry (2014)
Handbook of Pesticide Toxicology, Two-Volume Set: Principles and Agents, Volume 1, 1(2) (2001)
Handbook of Toxicology of Chemical Warfare Agents (2015)
Dietrich E Lorke et al.
Journal of applied toxicology : JAT, 39(11), 1506-1515 (2019-07-03)
Organophosphates, useful agents as pesticides, also represent a serious danger due to their high acute toxicity. There is indication that oximes, when administered before organophosphate exposure, can protect from these toxic effects. We have tested at equitoxic dosage (25% of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service