Skip to Content
Merck
All Photos(1)

Key Documents

74003

Supelco

(−)-Cotinine

analytical standard

Synonym(s):

(S)-1-Methyl-5-(3-pyridyl)-2-pyrrolidinone, S(−)-1-Methyl-5-(3-pyridyl)-2-pyrrolidone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O
CAS Number:
Molecular Weight:
176.22
Beilstein:
83099
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

optical activity

[α]/D -21±2°, c = 1 in ethanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

250 °C/150 mmHg (lit.)

mp

40-42 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

CN1[C@@H](CCC1=O)c2cccnc2

InChI

1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3/t9-/m0/s1

InChI key

UIKROCXWUNQSPJ-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Major metabolite of nicotine in humans.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mariana Belardinelli Rosa et al.
Acta odontologica Scandinavica, 78(4), 256-264 (2019-11-30)
Objective: The aim was to examine oral mechanical and gustatory sensitivities in adult smokers and to estimate salivary levels of cotinine by tobacco consumption. A total of 54 adults (20-45 years old; 28 males/26 females) were divided into two sex-paired groups:

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service