Skip to Content
Merck
All Photos(1)

Key Documents

73633

Supelco

Psoralen

analytical standard

Synonym(s):

6,7-Furanocoumarin, 7H-Furo[3,2-g]benzopyran-7-one, Ficusin, Furo[3,2-g]coumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H6O3
CAS Number:
Molecular Weight:
186.16
Beilstein:
152784
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O=C1Oc2cc3occc3cc2C=C1

InChI

1S/C11H6O3/c12-11-2-1-7-5-8-3-4-13-9(8)6-10(7)14-11/h1-6H

InChI key

ZCCUUQDIBDJBTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Photochemical reagent for the investigation of nucleic acid structure and function.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurelio Reyes et al.
Nucleic acids research, 41(11), 5837-5850 (2013-04-19)
The observation that long tracts of RNA are associated with replicating molecules of mitochondrial DNA (mtDNA) suggests that the mitochondrial genome of mammals is copied by an unorthodox mechanism. Here we show that these RNA-containing species are present in living
miR-TRAP: a benchtop chemical biology strategy to identify microRNA targets.
Huricha Baigude et al.
Angewandte Chemie (International ed. in English), 51(24), 5880-5883 (2012-05-09)
Biochemical aspects of psoralen photochemotherapy.
A C Moor et al.
Clinics in dermatology, 14(4), 353-365 (1996-07-01)
Maria Anokhina et al.
The EMBO journal, 32(21), 2804-2818 (2013-09-05)
Although U snRNAs play essential roles in splicing, little is known about the 3D arrangement of U2, U6, and U5 snRNAs and the pre-mRNA in active spliceosomes. To elucidate their relative spatial organization and dynamic rearrangement, we examined the RNA
Avvaru N Suhasini et al.
Molecular and cellular biology, 33(11), 2212-2227 (2013-03-27)
FANCJ mutations are linked to Fanconi anemia (FA) and increase breast cancer risk. FANCJ encodes a DNA helicase implicated in homologous recombination (HR) repair of double-strand breaks (DSBs) and interstrand cross-links (ICLs), but its mechanism of action is not well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service