Skip to Content
Merck
All Photos(1)

Key Documents

73257

Millipore

DNase ID Membrane

blue colored, sterile, diameter 70 mm, suitable for microbiology

Synonym(s):

Indicator membrane, ID Membranes

Sign Into View Organizational & Contract Pricing

Select a Size

25 DISCS
$41.50

$41.50

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request more information

Select a Size

Change View
25 DISCS
$41.50

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

$41.50

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request more information

membrane size

70 mm

sterility

sterile

form

disc

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Staphylococcus spp.

General description

DNase ID Membrane is a substrate-carrying ID membrane used for rapid identification of deoxyribonuclease (DNAse) activity of bacteria, especially for identification of pathogenic Staphylococci through characteristic color reactions. The membranes contain chromogenic substrates, which serve as the basis for microbe differentiation by color. The principle is based on an enzyme reaction in which specific DNase-producing Staphylococcus sp. will exhibit pink zones around blue color colonies.

Application

DNase ID Membrane is used for rapid identification of deoxyribonuclease (DNAse) activity of bacteria, especially for identification of pathogenic Staphylococci in water and food samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Analysis Note

Staphylococcus aureus (25923) - Pink zone around the colony, DNase positive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service