Skip to Content
Merck
All Photos(1)

Documents

72014

Supelco

Sodium tetrakis(4-fluorophenyl)borate dihydrate

Selectophore, ≥97.0%

Synonym(s):

Cesibor, Tetrakis(4-fluorophenyl)boron sodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(FC6H4)4BNa·2H2O
CAS Number:
Molecular Weight:
450.21
Beilstein:
4285808
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥97.0% (NT)
≥97.0%

form

solid

storage temp.

2-8°C

SMILES string

O.O.[Na+].Fc1ccc(cc1)[B-](c2ccc(F)cc2)(c3ccc(F)cc3)c4ccc(F)cc4

InChI

1S/C24H16BF4.Na.2H2O/c26-21-9-1-17(2-10-21)25(18-3-11-22(27)12-4-18,19-5-13-23(28)14-6-19)20-7-15-24(29)16-8-20;;;/h1-16H;;2*1H2/q-1;+1;;

InChI key

MSDGDEJOIBMWJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium tetrakis(4-fluorophenyl)borate dihydrate is sodium salt derivative of tetraphenylborate (TPB).
Visit our Sensor Applications portal to learn more.

Application

Anionic additive Sodium tetrakis(4-fluorophenyl)borate dihydrate has been used during the potentiometric measurement of ammonia and blood urea nitrogen (BUN) sensors.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Additive-salt effect on low detection limit and slope sensitivity in response of potassium-and sodium-selective neutral carrier based electrodes and their liquid-membrane based ion-sensitive field-effect transistor.
Wakida, Shin-ichi, et al.
Analytical Sciences, 15.1, 47-51 (1999)
Healthcare chip based on integrated electrochemical sensors used for clinical diagnostics of BUN.
Chang, Chia-Hsien, et al.
Jpn J. App. Phys., Part I, 45.5R, 4241-4241 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service