Skip to Content
Merck
All Photos(4)

Key Documents

71016BC

Millipore

GDS MPX Top 6 STEC

BioControl, Molecular based PCR test for the detection of the "Top Six" non-O157 Shiga Toxigenic E. coli (STEC) in food

Sign Into View Organizational & Contract Pricing

Select a Size

100 TEST
$1,530.00

$1,530.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
100 TEST
$1,530.00

About This Item

UNSPSC Code:
41106302
NACRES:
NB.61

$1,530.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Quality Level

shelf life

6‑12 mo.

manufacturer/tradename

BioControl Systems 71016-100

storage temp.

2-8°C

General description

GDS MPX Top 6 STEC is an automated nucleic acid amplification system for the detection of the “Top Six” non-O157 Shiga Toxigenic E. coli (STEC) in raw ground beef, raw beef trim, raw spinach, and raw mixed greens (baby lettuce and other young leafy green vegetables). The "Top Six" non-O157 STEC are defined as E. coli belonging to serogroups O103, O111, O121, O145, O26 or O45 that possess both the eae gene and at least one of the Shiga Toxin Genes stx1 or stx2. GDS MPX Top 6 STEC utilizes a proprietary IMS-based sample preparation procedure to isolate organisms belonging to 6 specific Top STEC O-serogroups (O103, O111, O121, O145, O26 and O45) prior to genetic analysis for the associated pathogenicity genes.

Features and Benefits

  • Following sample preparation with IMS against the Top 6 O groups, samples are analyzed with a single multiplex assay for the presence of (eae), stx1, and stx2
  • Single-step 10-18 h enrichment in mEHEC®, enrichment media common to all BioControl methods for E.coli O157:H7, Shiga Toxin genes, and Top STEC

Components

  • MPX Top STEC Amplification Tubes
  • Top 6 STEC Concentration Reagent
  • Resuspension Buffer Tq
  • Top STEC Wash Solution

Compatibility

BioControl GDS

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

mEHEC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service