Skip to Content
Merck
All Photos(1)

Key Documents

70310

Sigma-Aldrich

Sodium 1,3,(6,7)-naphthalenetrisulfonate tribasic hydrate

technical, ≥90.0% (T), mixture of isomers

Synonym(s):

Naphthalene-1,3,(6,7)-trisulfonic acid trisodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5Na3O9S3 · xH2O
CAS Number:
Molecular Weight:
434.31 (anhydrous basis)
Beilstein:
4637600
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90.0% (T)

composition

1,3,6-isomer, ~70%
1,3,7-isomer, ~30%

impurities

≤5% Na2SO4
≤6% water

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service