Skip to Content
Merck
All Photos(1)

Key Documents

70175

Sigma-Aldrich

Peptone from meat, enzymatic digest

suitable for microbiology

Synonym(s):

Peptone from bovine meat

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$81.60
500 G
$314.00

$81.60

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
100 G
$81.60
500 G
$314.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42040102
NACRES:
NA.74

$81.60

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


biological source

bovine skeletal muscle

form

powder

technique(s)

microbiological culture: suitable

impurities

≥10% total nitrogen (N)
≥3% amino N

ign. residue

≤12%

loss

≤6% loss on drying

pH

7.0±0.5 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear, slightly yellow

application(s)

food and beverages
microbiology

suitability

Clostridium spp.
Corynebacterium spp.
Salmonella spp.
Staphylococcus spp.

Looking for similar products? Visit Product Comparison Guide

General description

A specialized peptone prepared by the enzymatic digestion of selected fresh meat which can be used in media for the production of bacterial toxins from organisms such as Corynebacterium diphtheriae, Staphylococci, Clostridiae, and Salmonellae.

Application

Peptone from meat, enzymatic digest has been used:

  • to culture Candida albicans[1]
  • to study its effects on canonical and non-canonical receptors of glucagon-like peptide 1 (GLP1) in human colon tissue in different ex vivo models[2]
  • as a component of the nutrient broth (NB) to study its efficacy against pathogen resistance in grapevine plants[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carme Grau-Bové et al.
International journal of molecular sciences, 23(7) (2022-04-13)
GLP1 produced in the upper part of the gut is released after food intake and acts by activating insulin secretion, but the role of GLP1 in the colon, where it is predominantly produced, remains unknown. Here we characterized the apical
GSPE Pre-Treatment Exerts Long-Lasting Preventive Effects against Aging-Induced Changes in the Colonic Enterohormone Profile of Female Rats.
Migu??ns-G??mez, et al.
International Journal of Molecular Sciences, 24 (2023)
Martina Cappelletti et al.
Frontiers in plant science, 7, 1053-1053 (2016-08-04)
Protein derivatives and carbohydrates can stimulate plant growth, increase stress tolerance, and activate plant defense mechanisms. However, these molecules can also act as a nutritional substrate for microbial communities living on the plant phyllosphere and possibly affect their biocontrol activity
Olena P Ishchuk et al.
Antibiotics (Basel, Switzerland), 9(1) (2020-01-08)
The opportunistic human fungal pathogen Candida albicans relies on cell morphological transitions to develop biofilm and invade the host. In the current study, we developed new regulatory molecules, which inhibit the morphological transition of C. albicans from yeast-form cells to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service