69779
6′,7′-Epoxybergamottin
analytical standard
Synonym(s):
4-[[(2E)-5-(3,3-Dimethyl-2-oxiranyl)-3-methyl-2-penten-1-yl]oxy]-7H-furo[3,2-g][1]benzopyran-7-one, 5-(6′,7′-Epoxy)geranyloxypsoralen, Bergamottin 6′,7′-epoxide, Epoxybergamottin
About This Item
Recommended Products
grade
analytical standard
Assay
≥95.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
≤5.0% water
application(s)
food and beverages
format
neat
shipped in
wet ice
storage temp.
−20°C
SMILES string
O=C(O1)C=CC2=C1C=C(OC=C3)C3=C2OC/C=C(C)/CCC4C(C)(C)O4
InChI
1S/C21H22O5/c1-13(4-6-18-21(2,3)26-18)8-10-24-20-14-5-7-19(22)25-17(14)12-16-15(20)9-11-23-16/h5,7-9,11-12,18H,4,6,10H2,1-3H3/b13-8+
InChI key
OOKSPQLCQUBEKU-MDWZMJQESA-N
Looking for similar products? Visit Product Comparison Guide
Application
Recommended products
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service