Skip to Content
Merck
All Photos(1)

Key Documents

68923

Supelco

Esculetin

analytical standard

Synonym(s):

6,7-Dihydroxycoumarin, Cichorigenin, Esculetin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O4
CAS Number:
Molecular Weight:
178.14
Beilstein:
152788
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5% (HPLC)
97.5-102.5% (NT)

shelf life

limited shelf life, expiry date on the label

mp

271-273 °C (lit.)

format

neat

SMILES string

Oc1cc2OC(=O)C=Cc2cc1O

InChI

1S/C9H6O4/c10-6-3-5-1-2-9(12)13-8(5)4-7(6)11/h1-4,10-11H

InChI key

ILEDWLMCKZNDJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Esculetin is a derivative of coumarin, found in various plant species including Cichorium intybus (chicory), Bougainvillea spectabilis, Artemisia capillaris, etc and leaves of Citrus limonia (Rutaceae), Ceratostigma willmottianum, etc. It exhibits multiple pharmacological activities and is found to be a potent inhibitor of cyclooxygenase, 5-lipoxygenase, 12-lipoxygenase, catechol-O-methyl transferase, NADPH oxidase and xanthine oxidase enzymes.

Application

Esculetin may be used as an analytical reference standard for the determination of the analyte in biological and water samples by chromatography-based techniques. It may also be used as a standard in the quantification of the analyte in traditional herbal medicines by solid-phase extraction and micellar electrokinetic capillary methods, respectively.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esculetin prevents liver damage induced by paracetamol and CCl4
Gilani AH, et al.
Pharmacological Research, 37(1), 31-35 (1998)
Determination of polyphenol components in herbal medicines by micellar electrokinetic capillary chromatography with Tween 20
Wang XK, et al.
Talanta, 74(1), 1-6 (2007)
High performance liquid chromatographic determination of esculetin and daphnetin in urine and plasma
Egan DA, et al.
Chromatographia, 58(9-10), 649-652 (2003)
Inhibition of cell cycle progression in human leukemia HL-60 cells by esculetin
Wang CJ, et al.
Cancer Letters, 183(2), 163-168 (2002)
Solid-phase extraction of esculetin from the ash bark of Chinese traditional medicine by using molecularly imprinted polymers
Hu SG, et al.
Journal of Chromatography A, 1062(1), 31-37 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service