Skip to Content
Merck
All Photos(1)

Key Documents

68696

Supelco

Hexachlorobenzene-13C6

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$492.00

$492.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$492.00

About This Item

Linear Formula:
13C6Cl6
CAS Number:
Molecular Weight:
290.74
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

$492.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

isotopic purity

≥99 atom % 13C

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

mass shift

M+6

SMILES string

Cl[13c]1[13c](Cl)[13c](Cl)[13c](Cl)[13c](Cl)[13c]1Cl

InChI

1S/C6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

CKAPSXZOOQJIBF-IDEBNGHGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexachlorobenzene-13C6 is an isotope-labeled analog of the organochlorine pesticide, hexachlorobenzene.[1]

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The main advantage of using the IDMS technique is because isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analog, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genotoxicity of the organochlorine pesticides 1, 1-dichloro-2, 2-bis (p-chlorophenyl) ethylene (DDE) and hexachlorobenzene (HCB) in cultured human lymphocytes
Ennaceur S, et al.
Chemosphere, 71(7), 1335-1339 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service