Skip to Content
Merck
All Photos(1)

Key Documents

68633

Supelco

1,3-Dinonadecanoin

analytical standard

Synonym(s):

1,3-Dinonadecanoyl-rac-glycerol, 1,3-Dinonadecanoylglycerol, DG(19:0/0:0/19:0) [rac], Glyceryl dinonadecanoate

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$1,580.00

$1,580.00


Estimated to ship onMay 04, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$1,580.00

About This Item

Empirical Formula (Hill Notation):
C41H80O5
CAS Number:
Molecular Weight:
653.07
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$1,580.00


Estimated to ship onMay 04, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

ester

SMILES string

CCCCCCCCCCCCCCCCCCC(=O)OCC(O)COC(=O)CCCCCCCCCCCCCCCCCC

InChI

1S/C41H80O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-40(43)45-37-39(42)38-46-41(44)36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h39,42H,3-38H2,1-2H3

InChI key

VSZCJVIEISRCQF-UHFFFAOYSA-N

Application

1,3-Dinonadecanoin has been used as a lipid standard in the quantification of phospholipids from dairy products using solid-phase extraction followed by high-performance liquid chromatography (HPLC) with UV or evaporative light scattering detector (ELSD).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of phospholipids in dairy products by SPE/HPLC/ELSD.
Avalli A and Contarini G
Journal of Chromatography A, 1071(1-2), 185-190 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service