Skip to Content
Merck
All Photos(2)

Key Documents

68527

Supelco

Harpagoside

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30O11
CAS Number:
Molecular Weight:
494.49
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C[C@@]1(C[C@@H](O)[C@]2(O)C=CO[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@H]12)OC(=O)\C=C\c4ccccc4

InChI

1S/C24H30O11/c1-23(35-16(27)8-7-13-5-3-2-4-6-13)11-15(26)24(31)9-10-32-22(20(23)24)34-21-19(30)18(29)17(28)14(12-25)33-21/h2-10,14-15,17-22,25-26,28-31H,11-12H2,1H3/b8-7+/t14-,15-,17-,18+,19-,20-,21+,22+,23+,24-/m1/s1

InChI key

KVRQGMOSZKPBNS-FMHLWDFHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Harpagoside, a naturally occurring iridoid glycoside, is the main bioactive component of medicinal plants like Radix Scrophulariae, Scrophularia ningpoensis, Scrophularia buergeriana and Harpagophytum procumbens. It can exhibit a wide range of biological and pharmacological properties and hence used in the treatment of ailments like pharyngalgia, arthritis, rheumatism, tussis and constipation.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harpagoside: from Kalahari Desert to pharmacy shelf
Georgiev.IM, et al.
Phytochemistry, 92, 8-15 (2013)
Simultaneous determination of harpagoside and cinnamic acid in rat plasma by high-performance liquid chromatography: application to a pharmacokinetic study
Li P, et al.
Analytical and Bioanalytical Chemistry, 389, 2259-2264 (2007)
Nadia Romiti et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 16(12), 1095-1100 (2009-07-07)
Devil's Claw (Harpagophytum procumbens) a plant native to Southern Africa, has historically been used in traditional medicine to treat a wide range of diseases and currently is widely employed as anti-inflammatory and pain-relieving natural remedy in Europe and other parts
Shuting Yang et al.
Journal of agricultural and food chemistry, 59(5), 1612-1621 (2011-02-17)
Scrophularia ningpoensis Hemsl. is an important Chinese medicinal herb with a history of domestication of over 1000 years. Phytochemical variation of S. ningpoensis in response to environmental gradients remains an attractive topic with both practiceal and theoretical significances. In the
Corinna Boeckenholt et al.
Planta medica, 78(8), 761-766 (2012-03-14)
Silymarin and harpagoside are derived from drugs which are used for their protective effects against hepatotoxicity and inflammatory processes. Both are now investigated with respect to the respiratory tract. They were able to reduce the release of the inflammatory cytokine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service