Skip to Content
Merck
All Photos(1)

Key Documents

67884

Supelco

Fluorescein Sodium salt - CAPS solution

for fluorescence, ≥95.0% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$105.00

$105.00


Estimated to ship onMarch 26, 2025


Request a Bulk Order

Select a Size

Change View
10 ML
$105.00

About This Item

UNSPSC Code:
12190000
NACRES:
NA.21

$105.00


Estimated to ship onMarch 26, 2025


Request a Bulk Order

grade

for fluorescence

Quality Level

Assay

≥95.0% (HPLC)

concentration

1 mM fluorescein sodium salt in 20 mM CAPS, pH 10.0

λmax

490 nm±5 nm

fluorescence

λex 490 nm; λem 514 nm in 20 mM CAPS, pH 10.0

detection method

fluorometric

storage temp.

2-8°C

General description

recommended storage temperature 15 °C

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service