Skip to Content
Merck
All Photos(1)

Key Documents

67667

Supelco

Kynurenic acid

analytical standard

Synonym(s):

KYNA, 4-Hydroxyquinoline-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO3
CAS Number:
Molecular Weight:
189.17
Beilstein:
147451
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

275 °C (dec.) (lit.)

application(s)

clinical testing

SMILES string

OC(=O)c1cc(O)c2ccccc2n1

InChI

1S/C10H7NO3/c12-9-5-8(10(13)14)11-7-4-2-1-3-6(7)9/h1-5H,(H,11,12)(H,13,14)

InChI key

HCZHHEIFKROPDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kynurenic acid, a tryptophan metabolite, is a naturally occurring NMDA-receptor and an α7 nicotinic acetylcholine receptor antagonist in the human brain with preferential action at both of the glycine (Gly) binding sites of the N-methyl D-aspartate (NMDA) and α7 nicotinic acetylcholine receptors. This implies that the compound can have a physiological interaction with glutaminergic and cholinergic neurotransmission in the brain. Kynurenic acid is reported to be found in elevated levels in the cerebrospinal fluid of patients with schizophrenia. Its anticonvulsant and neuroprotective properties are experimentally reported.

Application

Kynurenic acid may be used as an analytical standard for the quantification of the analyte in serum samples, food and honeybee products using high-performance liquid chromatography with fluorescence detection.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

More rapid and sensitive method for simultaneous determination of tryptophan and kynurenic acid by HPLC
Lan-Gan P, et al.
Clinical Biochemistry, 42(4-5), 420-425 (2009)
Determination of kynurenic acid in human serum and its correlation with the concentration of certain amino acids
Fukushima T, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 377(1-2), 174-178 (2007)
Presence of kynurenic acid in food and honeybee products
Turski PM, et al.
Amino Acids, 36(1), 75-80 (2009)
Kynurenic acid levels are elevated in the cerebrospinal fluid of patients with schizophrenia
Erhardt S, et al.
Neuroscience Letters, 313(1-2), 96-98 (2001)
Kynurenic acid inhibits synaptic and acidic amino acid-induced responses in the rat hippocampus and spinal cord
Ganong HA, et al.
Brain Research, 273(1), 170-174 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service