Skip to Content
Merck
All Photos(3)

Key Documents

67416

Millipore

Acinetobacter Millichrome plus Agar

selective for bacteria: Acinetobacter spp, suitable for microbiology

Synonym(s):

Acinetobacter culture medium, Acinetobacter identification agar, Acinetobacter selective agar, AM plus Agar, Millichrome plus selective agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106204
NACRES:
NA.74
Pricing and availability is not currently available.

Agency

EP
USP

Quality Level

Sterilization Compatibility

dry heat compatible
compatible

form

dry powder

shelf life

limited shelf life, expiry date on the label

composition

Agar, 15.0 g/L
Chromogenic mix, 1.8 g/L
Peptone and yeast extract 12.0
Salts, 4.0 g/L

manufacturer/tradename

MilliChrome

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

solubility

deionized water: 32.8 g/L

application(s)

microbiology
pharmaceutical
water monitoring

shipped in

ambient

storage temp.

15-25°C

suitability

selective for bacteria: Acinetobacter MDR
selective for bacteria: Acinetobacter spp

General description

Acinetobacter Millichrome plus Agar is a selective and differential chromogenic culture medium, intended for use in the Acinetobacter and MDR Acinetobacter spp. The components of the medium are peptone and yeast extract, salts, chromogenic mix, and agar. Peptone and yeast extract provide nutrients for growth, salts help with osmotic balance and provide essential ions, the chromogenic mix contains substrates for color differentiation based on enzymatic activity, and agar is the solidifying agent. The medium is designed to selectively and differentially detect Acinetobacter, including multi-drug resistant strains (by supplementing with MDR Acinetobacter Millichrome plus Supplement ), and aid in the prevention and control of Acinetobacter in healthcare and pharma settings.

Application

Acinetobacter Millichrome plus Agar Base is used for the qualitative direct detection of colonization with Acinetobacter spp. to aid in the prevention and control of Acinetobacter, drug-susceptible or multi-drug resistant (MDR), in healthcare settings and pharma production. It can also be used in hygiene monitoring in the clinical and pharma environment with surface sampling.

Preparation Note

Step 1: Disperse slowly 32.8 g of powder base in 1 L of purified water. Add 4.0 mL of the liquid Acinetobacter Chromocult® plus Supplement into slurry. Stir until agar is well thickened. Heat and bring to boil (100 °C) while swirling or stirring regularly. DO NOT HEAT TO MORE THAN 100 °C. DO NOT AUTOCLAVE AT 121 °C. Warning: If using an autoclave, do so without pressure. Advice: in case of product samples containing a high load of Pseudomonas and/or Aeromonas, Cefsulodin can be added at 5 mg/L. Cool in a water bath to 45-50 °C, swirling or stirring gently. Step 2: (optional, If screening is focused on MDR Acinetobacter, add the MDR Selective supplement as following:): Rehydrate one vial with 5 mL of purified water. Add 5 mL of this solution to 1 L of the melted mix (step 1) at 45-50 °C. Stir well for homogenization Step 3: Pour into sterile Petri dishes. Let it solidify and dry. Warning: Slight variation of the media coloration after solidification can be observed, from yellowish to light orange without any impact on the media performance. Add 5 mL of this solution to the melted mix (step 1) at 45-50 °C. Stir well for homogenization.

Storage and Stability

  • Store prepared media below 8°C, protected from direct light (max. 1 month).
  • Store dehydrated powder, in a dry place, in tightly sealed containers at 2-25°C.

Analysis Note

Characteristic red color colonies indicate the Acinetobacter after 24 h at an optimal growth temperature of 37°C. At the same time, most other Gram-negative bacteria form blue or inhibited after incubation at the same temperature.

Legal Information

CHROMOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
MilliChrome is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service