Skip to Content
Merck
All Photos(1)

Key Documents

65950

Sigma-Aldrich

Bromomethane

purum, ampule, ≥99.5% (GC)

Synonym(s):

Methyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3Br
CAS Number:
Molecular Weight:
94.94
Beilstein:
1209223
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

vapor density

3.3 (20 °C, vs air)

vapor pressure

1420 mmHg ( 20 °C)

grade

purum

Assay

≥99.5% (GC)

autoignition temp.

996 °F

quality

ampule

expl. lim.

~16 %

bp

4 °C (lit.)

mp

−94 °C (lit.)

density

1.73 g/mL at 20 °C
3.3 g/mL at 25 °C (lit.)

SMILES string

O[C@H]1[C@@H](O)[C@H](N2C=NC(C(N)=O)=C2N)O[C@@H]1COP(O)(O)=O
CBr

InChI

1S/CH3Br/c1-2/h1H3

InChI key

GZUXJHMPEANEGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Sales restrictions may apply

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Muta. 2 - Ozone 1 - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

<-22.0 °F - liquid

Flash Point(C)

< -30 °C - liquid

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yulong Xu et al.
The Journal of chemical physics, 137(18), 184501-184501 (2012-11-21)
The bimolecular nucleophilic substitution (S(N)2) reaction of CH(3)Br and OH(-) in aqueous solution was investigated using a multilayered-representation quantum mechanical and molecular mechanics methodology. Reactant complex, transition state, and product complex are identified and characterized in aqueous solution. The potentials
MMWR. Morbidity and mortality weekly report, 60(27), 923-926 (2011-07-15)
Methyl bromide (MeBr) is a toxic gas used to fumigate agricultural fields and some produce. The U.S. Department of Agriculture (USDA) requires MeBr fumigation of grapes imported from Chile to prevent invasion by the Chilean false red mite, Brevipalpus chilensis.
Kathryn Hughes Barry et al.
Cancer causes & control : CCC, 23(6), 807-818 (2012-04-25)
Methyl bromide is a genotoxic soil fumigant with high acute toxicity, but unknown human carcinogenicity. Although many countries have reduced methyl bromide use because of its ozone depleting properties, some uses remain in the United States and other countries, warranting
Michael J Cox et al.
FEMS microbiology letters, 334(2), 111-118 (2012-06-26)
Methyl halides have a significant impact on atmospheric chemistry, particularly in the degradation of stratospheric ozone. Bacteria are known to contribute to the degradation of methyl halides in the oceans and marine bacteria capable of using methyl bromide and methyl
Khristy J Thompson et al.
Toxicological sciences : an official journal of the Society of Toxicology, 120(1), 163-172 (2010-12-24)
Manganese (Mn) is an essential nutrient with potential neurotoxic effects. Mn deposited in the nose is apparently transported to the brain through anterograde axonal transport, bypassing the blood-brain barrier. However, the role of the olfactory epithelial cells in Mn transport

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service