Skip to Content
Merck
All Photos(1)

Documents

64001BC

Millipore

MVP ICON® Positive Controls

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112601
NACRES:
NA.85

manufacturer/tradename

BioControl Systems 64001-25
MVP ICON®

Quality Level

General description

The MVP ICON ATP Positive Control provides a sample which produces a positive ATP result when assayed with the MVP ICON system.

Application

It is recommended that ATP positive controls be run every other day of use and incorporated into an overall Quality Control program which monitors and tracks the MVP ICON system results over an extended period of time.

Components

Twenty-five (25) sealed glass vials containing approximately 1 x 10-13 moles of lyophilized Adenosine triphosphate (ATP).

Suitability

ATP Testing, Food Processing, HACCP Monitoring, Sanitation

Principle

The detection of ATP through bioluminescence is a well-established and widely used indicator of surface hygiene and cleaning efficacy in food processing. Visual inspection is not an adequate critical control for surface hygiene and the lengthy time-to-result of microbiological test methods (from 2 - 7 days) make them ineffective for hygiene monitoring. ATP detection with the MVP ICON provides results in 10 seconds allowing for immediate "go/no-go" decisions to be made prior to the start of processing and allows for correction actions, when necessary, to be taken immediately. ATP is a molecule that is found in all living cells, including: bacteria, yeast, mold and all organic matter. After cleaning and/or sanitizing, food processing equipment and the plant environment is tested for ATP. If cleaning/sanitizing has been effective, ATP levels should be low.

Testing Method

ATP Bioluminescence

Legal Information

MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • Lightning MVP ICON ATP Positive Control

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service