Skip to Content
Merck
All Photos(1)

Key Documents

63910

Sigma-Aldrich

Mesitylene

puriss., ≥99.0% (GC)

Synonym(s):

1,3,5-Trimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CH3)3
CAS Number:
Molecular Weight:
120.19
Beilstein:
906806
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

4.1 (vs air)

vapor pressure

14 mmHg ( 55 °C)
2.49 mmHg ( 25 °C)

grade

puriss.

Assay

≥99.0% (GC)

autoignition temp.

1022 °F

expl. lim.

0.88-6.1 %, 100 °F

refractive index

n20/D 1.499 (lit.)
n20/D 1.499

bp

163-166 °C (lit.)

mp

−45 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)cc(C)c1

InChI

1S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6H,1-3H3

InChI key

AUHZEENZYGFFBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

122.0 °F

Flash Point(C)

50 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Sprung et al.
Journal of the American Chemical Society, 137(5), 1916-1928 (2015-01-16)
A detailed and systematic polarized confocal fluorescence microscopy investigation is presented on three batches of large coffin-shaped ZSM-5 crystals (i.e., parent, steamed at 500 °C, and steamed at 700 °C). In total, six laser lines of different wavelength in the
Alessandro Scarso et al.
Journal of the American Chemical Society, 126(41), 13512-13518 (2004-10-14)
The reversible encapsulation of a series of normal alkane guests in a cylindrical host was studied by NMR methods. For small hydrocarbons such as n-pentane or n-hexane, two guests enter the host, and they move freely within. With n-heptane no
Soon Bang Kang et al.
The Journal of organic chemistry, 72(15), 5724-5730 (2007-06-28)
Facile synthesis of C-4 aryl pyrimidinone nucleoside analogues from an easily prepared O4-arylsulfonate derivative of thymidine is reported. Two O4-arylsulfonylthymidine precursors, (4-methylphenyl)sulfonyl and (2,4,6-trimethylphenyl)sulfonyl, were prepared and analyzed for their stabilities. Of the two, the latter possessed suitable stability as
John W Colson et al.
Science (New York, N.Y.), 332(6026), 228-231 (2011-04-09)
Covalent organic frameworks (COFs), in which molecular building blocks form robust microporous networks, are usually synthesized as insoluble and unprocessable powders. We have grown two-dimensional (2D) COF films on single-layer graphene (SLG) under operationally simple solvothermal conditions. The layered films
Natalie D Coombs et al.
Dalton transactions (Cambridge, England : 2003), (3)(3), 332-337 (2008-04-17)
Sterically encumbered amido ligands based on a 1,8-diarylcarbazol-9-yl backbone have been investigated as electronically distinct alternatives to the widely-used terphenyl ligand class in the stabilization of low-coordinate metal complexes, and structurally characterized for the first time. While 1,8-diphenylcarbazol-9-yl derivatives are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service