Skip to Content
Merck
All Photos(1)

Key Documents

63765

Sigma-Aldrich

Sodium 3-mercapto-1-propanesulfonate

technical, ~80% (RT)

Synonym(s):

3-Mercapto-1-propanesulfonic acid sodium salt, MPS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2CH2SO3Na
CAS Number:
Molecular Weight:
178.21
Beilstein:
5362242
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

~80% (RT)

form

solid

mp

~220 °C (dec.) (lit.)

SMILES string

[Na+].[O-]S(=O)(=O)CCCS

InChI

1S/C3H8O3S2.Na/c4-8(5,6)3-1-2-7;/h7H,1-3H2,(H,4,5,6);/q;+1/p-1

InChI key

FRTIVUOKBXDGPD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium 3-mercapto-1-propanesulfonate can also be involved in the synthesis of rhodium nanoparticles via liquid phase reduction method.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

"Highly dispersed noble-metal/chromia (core/shell) nanoparticles as efficient hydrogen evolution promoters for photocatalytic overall water splitting under visible light"
Sakamoto N, et al.
Nanoscale, 1(01), 106- 109 (2009)
S Q Wong et al.
British journal of cancer, 112(8), 1411-1420 (2015-03-06)
Recent discoveries in cancer research have revealed a plethora of clinically actionable mutations that provide therapeutic, prognostic and predictive benefit to patients. The feasibility of screening mutations as part of the routine clinical care of patients remains relatively unexplored as
Brener R C Vale et al.
Scientific reports, 9(1), 8332-8332 (2019-06-07)
CdTe/CdS core/shell quantum dots (QDs) are formed in aqueous synthesis via the partial decomposition of hydrophilic thiols, used as surface ligands. In this work, we investigate the influence of the chemical nature (functional group and chain length) of the used
Elsa G Shapiro et al.
Molecular genetics and metabolism, 114(4), 594-598 (2015-03-17)
The Sanfilippo Behavior Rating Scale (SBRS), a 68 item questionnaire, has been developed to assess the behavioral phenotype of children with Sanfilippo syndrome and its progression over time. Fifteen scales rate orality, movement/activity, attention/self-control, emotional function including anger and fear
Andrew J Foy et al.
JAMA internal medicine, 175(3), 428-436 (2015-01-27)
Patients presenting to the emergency department (ED) with chest pain whose evaluation for ischemia demonstrates no abnormalities receive further functional or anatomical studies for coronary artery disease; however, comparative evidence for the various strategies is lacking and multiple testing options

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service