Skip to Content
Merck
All Photos(1)

Documents

63177

Sigma-Aldrich

6-Maleimidohexanoic acid N-hydroxysuccinimide ester

≥98.0% (HPLC)

Synonym(s):

6-Maleimidocaproic acid N-succinimidyl ester, N-(ε-Maleimidocaproyloxy)succinimide, N-Succinimidyl 6-maleimidocaproate, EMCS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O6
CAS Number:
Molecular Weight:
308.29
Beilstein:
1499815
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.0% (HPLC)

form

solid

reaction suitability

reagent type: linker

mp

67-73 °C
70-73 °C (lit.)

functional group

NHS ester

storage temp.

−20°C

SMILES string

O=C(ON(C(CC1)=O)C1=O)CCCCCN2C(C=CC2=O)=O

InChI

1S/C14H16N2O6/c17-10-5-6-11(18)15(10)9-3-1-2-4-14(21)22-16-12(19)7-8-13(16)20/h5-6H,1-4,7-9H2

InChI key

VLARLSIGSPVYHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Maleimidohexanoic acid N-hydroxysuccinimide ester is a heterobifunctional cross-linking reagent with amine and sulfhydryl reactivity. Typically, coupled initially to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). Second coupling specific for molecules containing free sulfhydryl by thioether linkage buffered at pH 6.8 (6.5-7.0). Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Reagent for cross-linking and immobilisation of proteins

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T. Kitagawa
Enzyme Immunoassay, 81-81 (1981)
Y Nakano et al.
International archives of allergy and immunology, 120(3), 199-208 (1999-12-11)
We have previously reported that ovalbumin (OVA) coupled with liposome via glutaraldehyde (GA) induced OVA-specific- and IgE-selective unresponsiveness in mice. In this study, OVA-liposome conjugates were made using four different coupling protocols: via GA, N-(6-maleimidocaproyloxy) succinimide (EMCS), disuccinimidyl suberate (DSS)
N J Maeji et al.
Journal of immunological methods, 146(1), 83-90 (1992-01-21)
Recently, the multipin approach for simultaneous multiple peptide synthesis was applied to the analysis of T cell determinants by using a novel cleavage method (Maeji et al., 1990). A diketopiperazine forming linker allowed cleavage of peptides into aqueous buffer which
Mitsuko Maeda et al.
Bioorganic & medicinal chemistry letters, 15(3), 621-624 (2005-01-25)
The adenovirus vector is a promising carrier for the efficient transfer of genes into cells via the coxackie-adenovirus receptor (CAR) and integrins (alphavbeta3 and alphavbeta5). The clinical use of the adenovirus vector remains problematic however. Successful administration of this vector
Shinya Kida et al.
Chemical & pharmaceutical bulletin, 55(4), 685-687 (2007-04-06)
6-maleimidohexanoic acid N-hydroxysuccinimide ester has been used widely for preparation of enzyme immunoconjugates as a unique heterobifunctional cross-linking reagent. Its heterobifunctional reactivity is good, but its ester portion hydrolyzes easily in the presence of water. Several 6-maleimidohexanoic acid active esters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service