Skip to Content
Merck
All Photos(1)

Documents

62559

Supelco

Lithium ionophore II

Selectophore, function tested

Synonym(s):

N,N,N′,N′-Tetraisobutyl-cis-cyclohexane-1,2-dicarboxamide, ETH 1644

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H46N2O2
CAS Number:
Molecular Weight:
394.63
Beilstein:
6428727
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

Quality Level

quality

function tested

SMILES string

CC(C)CN(CC(C)C)C(=O)[C@H]1CCCC[C@H]1C(=O)N(CC(C)C)CC(C)C

InChI

1S/C24H46N2O2/c1-17(2)13-25(14-18(3)4)23(27)21-11-9-10-12-22(21)24(28)26(15-19(5)6)16-20(7)8/h17-22H,9-16H2,1-8H3/t21-,22+

InChI key

GPFAWYFTCKIPRA-SZPZYZBQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Neutral ionophore for a lithium ion-selective solvent polymeric membrane;

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Zeevi et al.
The Journal of membrane biology, 86(1), 61-67 (1985-01-01)
The neutral noncyclic, lithium-selective ionophore ETH1644, which is structurally different from previously available ionophores of this type, is a selective carrier of Li+ in lipid bilayer membranes of various lipid composition. The ionophore forms a 2:1 carrier/cation complex, and the
A.F. Zhukow et al.
Analytica Chimica Acta, 131, 177-177 (1981)
Erne, D., et al.
Helvetica Chimica Acta, 65, 538-538 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service