Skip to Content
Merck
All Photos(1)

Key Documents

61895

Sigma-Aldrich

[Leu13, Glu14]Motilin, porcine

PeptiSelect

Synonym(s):

[13-Leucine, 14-desamido]Motilin (pig), [13-Leucine, 14-glutamic acid]Motilin, porcine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C121H189N33O36
CAS Number:
Molecular Weight:
2681.99
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

PeptiSelect

shipped in

wet ice

storage temp.

−20°C

Antibody Adsorbant

H-Phe-Val-Pro-Ile-Phe-Thr-Tyr-Gly-Glu-Leu-Gln-Arg-Leu-Glu-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln-OH

Other Notes

Reviews on motilin[1][2][3][4]
Sales restrictions may apply

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V. Mutt
Brain Peptides, 871-871 (1983)
S.R. Bloom et al.
Hormones in Blood, 321-321 (1979)
G. Bertaccini
Handbook of Experimental Pharmacology, 59, 85-85 (1982)
J E Fox
Life sciences, 35(7), 695-706 (1984-08-13)
Motilin isolated in 1971 from the porcine gastrointestinal tract and localized there to endocrine cells, now appears to have a CNS neural origin by RIA and immunohistochemistry. In most species motilin releases neurotransmitters in the CNS to both increase and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service