Skip to Content
Merck
All Photos(1)

Key Documents

61169

Sigma-Aldrich

Copper(I) chloride

purum, ≥97.0% (RT)

Synonym(s):

Copper monochloride, Cuprous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl
CAS Number:
Molecular Weight:
99.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

vapor pressure

1.3 mmHg ( 546 °C)

grade

purum

Assay

≥97.0% (RT)

form

powder

reaction suitability

reagent type: catalyst
core: copper

bp

1490 °C (lit.)

mp

430 °C (lit.)

solubility

dimethyl sulfide: partially soluble(lit.)
water: insoluble(lit.)

SMILES string

Cl[Cu]

InChI

1S/ClH.Cu/h1H;/q;+1/p-1

InChI key

OXBLHERUFWYNTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Shows unique character as an initiator of radical reactions such as the hydrostannation of α,β-unsaturated ketones.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper (I) Chloride.
Bertz SH, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2007)
Experimental study of copper (I) chloride complexing in hydrothermal solutions at 40 to 300 ?C and saturated water vapor pressure.
Xiao Z, et al.
Geochimica et Cosmochimica Acta, 62(17), 2949-2964 (1998)
Copper (I) chloride-mediated intramolecular coupling of vinyltrimethylstannane and vinyl halide functions.
Piers E and Wong T.
The Journal of Organic Chemistry, 58(14), 3609-3610 (1993)
Xiaoliang Xu et al.
Organic letters, 12(5), 897-899 (2010-02-04)
Promoted by CuCl/CCl(4), a variety of sulfonyl azides and tertiary amines were successfully coupled to give sulfonyl amidine derivatives in good to excellent yields. A possible mechanism for this reaction is discussed.
Takayoshi Arai et al.
The Journal of organic chemistry, 73(13), 4903-4906 (2008-06-03)
A catalytic asymmetric Henry reaction has been developed with use of a sulfonyldiamine-CuCl complex as a catalyst. A series of new binaphthyl-containing sulfonyldiamine ligands (2a-h) were readily synthesized in two steps starting from commercially available chiral 1,2-diamines. The (R,R)-diamine-(R)-binaphthyl ligand

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service