Skip to Content
Merck
All Photos(1)

Key Documents

61165

Sigma-Aldrich

Copper(II) bromide

purum, ≥99.0% (RT)

Synonym(s):

Cupric bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuBr2
CAS Number:
Molecular Weight:
223.35
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

purum

Assay

≥99.0% (RT)

form

solid

reaction suitability

reagent type: catalyst
core: copper

mp

498 °C (lit.)

density

4.77 g/mL at 25 °C (lit.)

SMILES string

Br[Cu]Br

InChI

1S/2BrH.Cu/h2*1H;/q;;+2/p-2

InChI key

QTMDXZNDVAMKGV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shengqing Ye et al.
Chemical communications (Cambridge, England), (36)(36), 5406-5408 (2009-09-03)
Palladium-catalyzed tandem reactions of 2-alkenylphenyl-acetylenes with CuCl2 or CuBr2 afforded 3-chloro- or 3-bromo-1-methyleneindenes in good yields; these compounds could be further elaborated via palladium-catalyzed coupling reactions.
Robin B Bedford et al.
Dalton transactions (Cambridge, England : 2003), 39(43), 10464-10472 (2010-10-12)
The solvent-free, palladium-catalysed reaction of anilides with CuCl(2) in the presence or absence of copper acetate yields ortho-chlorinated anilides in good to excellent yields, even on a large scale (100 mmol). By contrast, the equivalent reactions with copper bromide, either
Yi-Si Feng et al.
Organic letters, 15(7), 1472-1475 (2013-03-12)
Cu-catalyzed decarboxylative alkynylation of quaternary α-cyano acetate salts with alkynyl bromides and alkynyl chlorides is described. This new reaction can be used for preparing functionalized butynenitrile derivatives.
Fei Yu et al.
The Journal of organic chemistry, 74(3), 1130-1134 (2009-01-07)
CuX(2)-mediated (X = Cl, Br) halolactonization of monoesters of 1,2-allenyl phosphonic acids is presented. The reaction proceeded smoothly under the mild condition for differently substituted allenic substrates giving the 4-halo-2,5-dihydro[1,2]oxaphosphole 2-oxides in good yields. The Suzuki cross-coupling reaction of these
Hong-Ying Niu et al.
Organic & biomolecular chemistry, 9(14), 5039-5042 (2011-06-11)
CuBr was found to be an efficient catalyst for the C-N cross coupling reaction of purine and diaryliodonium salts. 9-Arylpurines were synthesized in excellent yields with short reaction times (2.5 h). The method represents an alternative to the synthesis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service