Skip to Content
Merck
All Photos(1)

Key Documents

61020BC

Millipore

Assurance® GDS Top STEC Wash Solution PCR Reagent

For use with Assurance® GDS Top STEC

Synonym(s):

Wash solution for PCR Sample Prep

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 250 ML
$151.00

$151.00


Estimated to ship onMay 18, 2025


Request more information

Select a Size

Change View
4 X 250 ML
$151.00

About This Item

UNSPSC Code:
41106306
NACRES:
NA.85

$151.00


Estimated to ship onMay 18, 2025


Request more information

Product Name

GDS Top STEC Wash Solution Kit, BioControl, For use with GDS Top STEC

Quality Level

form

liquid

shelf life

6‑12 mo.

packaging

bottle of 4 × 250 mL each

manufacturer/tradename

BioControl Systems 61020-100

storage condition

(No metal containers.

Tightly closed.)

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

storage temp.

2-8°C

suitability

Escherichia coli (shiga toxigenic)

General description

GDS Top STEC Wash Solution Kit is recommended to use with Assurance® GDS Top STEC kits. Assurance® GDS Top STEC assays are automated nucleic acid amplification systems for the detection of Shiga toxigenic E. coli.

Application

GDS Top STEC Wash Solution Kit is used during the sample preparation for detection of Shiga toxigenic E. coli in foods and ingredients using the Assurance® GDS Top STEC assays.

Components

Four bottles, 250 mL each

Compatibility

BioControl GDS

Testing Method

PCR

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
BioControl is a registered trademark of BioControl Systems, Inc.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service