Skip to Content
Merck
All Photos(1)

Key Documents

60730

Sigma-Aldrich

Khellin

for microscopy

Synonym(s):

4,9-Dimethoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$591.00

$591.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$591.00

About This Item

Empirical Formula (Hill Notation):
C14H12O5
CAS Number:
Molecular Weight:
260.24
Beilstein:
263185
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

$591.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

grade

for microscopy

Quality Level

Assay

≥98.0% (HPLC)

form

powder

bp

180-200 °C/0.05 mmHg (lit.)

mp

150-154 °C (lit.)

suitability

suitable for microscopy

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

COc1c2OC(C)=CC(=O)c2c(OC)c3ccoc13

InChI

1S/C14H12O5/c1-7-6-9(15)10-11(16-2)8-4-5-18-12(8)14(17-3)13(10)19-7/h4-6H,1-3H3

InChI key

HSMPDPBYAYSOBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nishit S Patel et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 38(3), 381-391 (2012-02-01)
Vitiligo is an acquired multifocal and polygenic dyschromia that affects 1% to 3% of the world and presents as multiple depigmented macules and patches. Traditionally, the treatment of vitiligo has focused on pharmacologic interventions, but nearly half of all treated
Sally S El-Nakkady et al.
Acta poloniae pharmaceutica, 69(4), 645-655 (2012-08-11)
Bromination of visnaginone (1) yielded the dibromo derivative (2), which upon methylation with methyl iodide gave 1-(2,7-dibromo-4,6-dimethoxybenzofuran-5-yl) ethanone (3). Compound (3) reacted with dimethylformamide dimethylacetal to give (4). The reaction of (3) with aromatic aldehydes namely (vanillin, benzaldehyde and 3-anisaldehyde)
Rosita Saraceno et al.
Dermatologic therapy, 22(4), 391-394 (2009-07-08)
Vitiligo is an acquired depigmentation disorder affecting 1-4% of the world's population. Conventional therapies include steroids, photosensitive topical agents, surgical treatments, and phototherapy. The aim of the study was to evaluate the efficacy of monochromatic excimer light 308 nm (MEL)
P Vanachayangkul et al.
Urological research, 39(3), 189-195 (2010-11-12)
In Egypt, teas prepared from the fruits of Ammi visnaga L. (syn. "Khella") are traditionally used by patients with urolithiasis. The aim of this study was to evaluate whether oral administration of an aqueous extract prepared from the fruits of
Karin G Haug et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 45(1-2), 79-89 (2011-11-17)
Ammi visnaga L. (syn. Khella, Apiaceae) preparations have traditionally been used in the Middle East for the treatment of kidney stone disease. Visnagin, a furanocoumarin derivative, is one of the main compounds of Ammi visnaga with potential effects on kidney

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service