Skip to Content
Merck

59532

Sigma-Aldrich

(4R,5R)-2,2-Dimethyl-α,α,α′,α′-tetraphenyldioxolane-4,5-dimethanol

purum, ≥97.0% (sum of enantiomers, HPLC)

Synonym(s):

(−)-2,3-O-Isopropylidene-1,1,4,4-tetraphenyl-L-threitol, (4R,5R)-4,5-Bis(diphenylhydroxymethyl)-2,2-dimethyldioxolane, 1,1,4,4-Tetraphenyl-2,3-O-isopropylidene-L-threitol, TADDOL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H30O4
CAS Number:
Molecular Weight:
466.57
Beilstein:
3657855
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Pricing and availability is not currently available.

grade

purum

Assay

≥97.0% (sum of enantiomers, HPLC)

optical activity

[α]20/D −67±2°, c = 1% in chloroform

mp

192-195 °C

SMILES string

CC1(C)O[C@H]([C@@H](O1)C(O)(c2ccccc2)c3ccccc3)C(O)(c4ccccc4)c5ccccc5

Looking for similar products? Visit Product Comparison Guide

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service