Skip to Content
Merck
All Photos(1)

Key Documents

59438

Sigma-Aldrich

N-Isopropylhydroxylamine oxalate salt

purum, ≥95.0% (NT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHNHOH]2 · H2C2O4
CAS Number:
Molecular Weight:
240.25
Beilstein:
605272
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

purum

Assay

≥95.0% (NT)

mp

159-160 °C (dec.)

SMILES string

CC(C)NO.CC(C)NO.OC(=O)C(O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service