Skip to Content
Merck
All Photos(1)

Key Documents

58320

Sigma-Aldrich

D-(−)-Isoascorbic acid

purum, ≥99.0% (RT)

Synonym(s):

D-erythro-Hex-2-enoic acid γ-lactone, D-Araboascorbic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84271
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

purum

Assay

≥99.0% (RT)

optical activity

[α]20/D −16.0±1.5°, c = 1% in H2O

solubility

H2O: 0.1 g/mL, clear, colorless to very faintly yellow

SMILES string

OC[C@@H](O)[C@H]1OC(=O)C(O)=C1O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5-/m1/s1

InChI key

CIWBSHSKHKDKBQ-DUZGATOHSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Starting material for the synthesis of chiral building blocks

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.A. Vekemans et al.
Rec. Trav. Chim., 104, 266-266 (1985)
The Journal of Organic Chemistry, 52, 1093-1093 (1987)
Anna Lewinska et al.
Fungal genetics and biology : FG & B, 45(8), 1182-1187 (2008-07-11)
Since the double Deltagrx1Deltagrx2 mutant is hypersensitive to selenite we decided to evaluate mechanisms underlying this phenomenon and establish the roles of other components of yeast glutaredoxin system, in particular glutaredoxin 5 in the selenite resistance. We found elevation in
Vincenzo Carbone et al.
Acta crystallographica. Section D, Biological crystallography, 64(Pt 5), 532-542 (2008-05-06)
Mammalian dimeric dihydrodiol dehydrogenase (DD) is identical to NADP+-dependent D-xylose dehydrogenase. A recent investigation showed that the three-dimensional structure of monkey DD is similar to those of prokaryotic NADP(H)-dependent glucose-fructose oxidoreductase (GFO) and 1,5-anhydro-D-fructose reductase (AFR); however, it differs in
Liyun Wang et al.
Biochemical and biophysical research communications, 384(2), 210-214 (2009-04-29)
We recently identified a microbial conversion of L-ascorbic acid (AsA) to L-erythroascorbic acid (eAsA), a five-carbon analog of AsA. In this paper, we show that ubiquitin plays a crucial role in this process. Based on an assay that determined AsA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service