Skip to Content
Merck
All Photos(1)

Key Documents

58279

Sigma-Aldrich

Isatoic anhydride

technical, ≥94% (HPLC)

Synonym(s):

3,1-Benzoxazine-2,4(1H)-dione, Anthranilic acid N-carboxylic acid anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5NO3
CAS Number:
Molecular Weight:
163.13
Beilstein:
136786
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor density

5.6 (vs air)

grade

technical

Assay

≥94% (HPLC)

mp

233 °C (dec.) (lit.)

SMILES string

O=C1Nc2ccccc2C(=O)O1

InChI

1S/C8H5NO3/c10-7-5-3-1-2-4-6(5)9-8(11)12-7/h1-4H,(H,9,11)

InChI key

TXJUTRJFNRYTHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Review; Reagent for 2-aminobenzoylations

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.C. Venuti
Synthesis, 266-266 (1982)
G.M. Coppola
Synthesis, 505-505 (1980)
M G Page
The Biochemical journal, 295 ( Pt 1), 295-304 (1993-10-01)
Class C beta-lactamases from Pseudomonas aeruginosa and several species of the Enterobacteriaceae have been observed to undergo a rapid burst in hydrolysis of beta-lactam antibiotics before relaxation to a steady-state rate of hydrolysis. The amplitude of the burst corresponds to
Christine Hiu-Tung Chen et al.
Molecular diversity, 9(4), 353-359 (2005-11-29)
Reactions using fluorous reagents and scavengers are compared side-by-side with their solid-supported counterparts. Fluorous triphenylphosphine is used in the bromination reaction of alcohols, fluorous thiol is used as an electrophile scavenger for alpha-bromoketones, fluorous isatoic anhydride is used as a
J E Churchich
Analytical biochemistry, 213(2), 229-233 (1993-09-01)
Isatoic anhydride reacts with nucleophile groups of proteins to yield o-aminobenzoyl protein conjugates. The fluorescence emitted by the chromophore decays in a multiexponential manner with average fluorescence lifetimes ranging from 9.9 to 10.7 ns. The steady emission anisotropy, measured upon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service