56865
Ammonium trifluoroacetate
suitable for HPLC, 99.0-101.0% (NT)
Synonym(s):
Trifluoroacetic acid ammonium salt
About This Item
Recommended Products
Assay
99.0-101.0% (NT)
form
solid
technique(s)
HPLC: suitable
pH
5-6.5
mp
123-125 °C (lit.)
solubility
H2O: 10%, clear, colorless
λ
10 % in H2O
UV absorption
λ: 260 nm Amax: ≤0.05
λ: 270 nm Amax: ≤0.04
λ: 280 nm Amax: ≤0.03
λ: 290 nm Amax: ≤0.02
λ: 500 nm Amax: ≤0.02
SMILES string
N.OC(=O)C(F)(F)F
InChI
1S/C2HF3O2.H3N/c3-2(4,5)1(6)7;/h(H,6,7);1H3
InChI key
YCNIBOIOWCTRCL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Specific-Ion Effects Unveil a Route for Modulating Oxidatively Triggered Acid Systems: The research utilizes ammonium trifluoroacetate to explore specific-ion effects in acid systems for reservoir applications. This application is vital for industrial processes, particularly in the optimization of resource extraction technologies (Cairns et al., 2022).
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Polar graphitic carbon enables UHPLC separation of highly polar pesticides, herbicides, and Vitamin D metabolites, advancing liquid chromatography capabilities.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service