Skip to Content
Merck
All Photos(1)

Key Documents

56694

Sigma-Aldrich

Acid Alcohol solution

for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
MA.02

biological source

synthetic

Quality Level

grade

for microscopy

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | staining: suitable

color

colorless

application(s)

hematology
histology

storage temp.

room temp

suitability

mycobacteria

General description

This acid alcohol is part of Flourecent Stain Kit for Mycobacteria (Cat. No. 05151). It is used for decolorising.

Application

Acid Alcohol solution has been used for washing RNA plaques isolated from intraoperative tumor specimens for gene mRNA expression.[1]

Components

Composition:
75 % aqueous Ethanol 99.5 mL, Sodium chloride 0.5 g, Hydrochloric acid 0.5 g

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Correlation study between helicobacter pylori infection and tumor malignancy in patients with gastric cancer
Chen XB
FABAD Journal of Pharmaceutical Sciences, 23(4), 48-51 (2017)
Jemma Victoria Walker et al.
Nature communications, 10(1), 3596-3596 (2019-08-11)
Stem cells (SCs) receive inductive cues from the surrounding microenvironment and cells. Limited molecular evidence has connected tissue-specific mesenchymal stem cells (MSCs) with mesenchymal transit amplifying cells (MTACs). Using mouse incisor as the model, we discover a population of MSCs

Articles

Identifying Mycobacterium species crucial due to varying clinical significance; some pathogenic, others not.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service