Skip to Content
Merck
All Photos(2)

Key Documents

56448

Supelco

Xanthotoxin

analytical standard

Synonym(s):

8-Methoxypsoralen, 8-MOP, 9-Methoxyfuro[3,2-g][1]benzopyran-7-one, Ammoidin, Methoxsalen, Xanthotoxin

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$171.00

$171.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$171.00

About This Item

Empirical Formula (Hill Notation):
C12H8O4
CAS Number:
Molecular Weight:
216.19
Beilstein:
196453
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$171.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

148-150 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

COc1c2OC(=O)C=Cc2cc3ccoc13

InChI

1S/C12H8O4/c1-14-12-10-8(4-5-15-10)6-7-2-3-9(13)16-11(7)12/h2-6H,1H3

InChI key

QXKHYNVANLEOEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xanthotoxin is an allelochemical or a phototoxic allomone mainly found in host plants of black swallowtail.[1] It can have the tendency to induce tolerance towards Α-cypermethrin, a pyrethroid insecticide used for insects like Helicoverpa zea.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

8-methoxypsoralen (8-MOP) plus ultraviolet A (UVA) irradiation induces monoadducts and interstrand cross-links in DNA and therefore can be used to study DNA repair and recombination mechanisms. Cultured normal human melanocytes treated with 8-methoxypsoralen and irradiated with ultraviolet A (UVA) formed 8-methoxypsoralen-phospholipid photoadducts that could be substituted for diacylglycerol to activate protein kinase C in a cell-free system. 8-MOP is an inactivator of purified reconstituted cytochrome P450. It also inhibits substance P-induced histamine release from substance P-activated rat peritoneal mast cells by suppressing the rise in [Ca2+].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: angelica carum pimpinella

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Molecular cloning and expression of CYP6B8: a xanthotoxin-inducible cytochrome P450 cDNA from Helicoverpa zea
Li X, et al.
Insect Biochemistry and Molecular Biology, 30, 75-84 (2000)
Induction of cytochrome P450-mediated detoxification of xanthotoxin in the black swallowtail
Cohen.BM, et al.
Journal of Chemical Ecology, 15, 2347-2355 (1989)
Katarzyna Borowska et al.
International journal of pharmaceutics, 426(1-2), 280-283 (2012-02-09)
In the present study we have assessed the ability of (PAMAM) dendrimers G3 and G4 to facilitate transdermal delivery of 8-methoxypsoralen (8-MOP) in vivo. In vitro study using Franz diffusion cell revealed an enhanced transdermal flux for 8-MOP in complex
Shruti Chowdhari et al.
Journal of cellular physiology, 229(11), 1630-1638 (2014-03-13)
Psoriasis is a chronic inflammatory skin disorder mediated by cross-talk occurring between epidermal keratinocytes, dermal vascular cells and immunocytes. Literature reveals that Signal transducer and activator of transcription 3 (STAT3), a protein involved in transmitting extracellular signals to the nucleus
Y Tokura
Journal of dermatological science, 19(2), 114-122 (1999-03-31)
Modulatory effects on cytokine expression of 8-MOP in conjunction with UVA have been investigated in different systems using different cell types, including keratinocytes, lymphocytes, monocytes and endothelial cells. The vast majority of data have exhibited reduced production of cytokines in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service