55842
Reserpine solution
1 pg/μL in methanol: water (1:1)
About This Item
Recommended Products
Quality Level
concentration
1 pg/μL in methanol: water (1:1)
technique(s)
HPLC: suitable
LC/MS: suitable
density
0.925-0.929 g/mL at 20 °C
suitability
corresponds for LC-MS
InChI
1S/C33H40N2O9/c1-38-19-7-8-20-21-9-10-35-16-18-13-27(44-32(36)17-11-25(39-2)30(41-4)26(12-17)40-3)31(42-5)28(33(37)43-6)22(18)15-24(35)29(21)34-23(20)14-19/h7-8,11-12,14,18,22,24,27-28,31,34H,9-10,13,15-16H2,1-6H3/t18-,22+,24-,27-,28+,31+/m1/s1
InChI key
QEVHRUUCFGRFIF-MDEJGZGSSA-N
Looking for similar products? Visit Product Comparison Guide
General description
- Extensive QC testing ensures the highest specifications.
- Optimized to improve ionization and resolution.
- Extremely low levels of inorganic and organic impurities.
- Manufactured specifically for accurate and fast LC-MS.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1
Target Organs
Eyes
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
75.2 °F
Flash Point(C)
24 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service