Skip to Content
Merck
All Photos(1)

Documents

55795

Supelco

Acetaldehyde diethyl acetal

analytical standard

Synonym(s):

1,1-Diethoxyethane, Acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OC2H5)2
CAS Number:
Molecular Weight:
118.17
Beilstein:
1098310
EC Number:
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.1 (vs air)

vapor pressure

20 mmHg ( 20 °C)
8 mmHg ( 8 °C)

Assay

≥98.0% (GC)

autoignition temp.

446 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

10.4 %

refractive index

n20/D 1.379-1.383 (lit.)

bp

102 °C (lit.)

density

0.831 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCOC(C)OCC

InChI

1S/C6H14O2/c1-4-7-6(3)8-5-2/h6H,4-5H2,1-3H3

InChI key

DHKHKXVYLBGOIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetaldehyde diethyl acetal is one of the precursors utilized for the synthesis of fragrances and pharmaceutical products.

Application

Acetaldehyde diethyl acetal may be used as a reference standard for the determination of acetaldehyde diethyl acetal in:
  • Wine vinegars by headspace sorptive extraction−thermal desorption−gas chromatography−mass spectrometry (HSSE−TD−GC−MS).
  • Distilled spirits by GC with flame ionization detection (FID) as well as GC-FID-MS.
  • Organic red wines derived from Saccharomyces cerevisiae strains by GC-FID as well as HSSE-GC−MS.
  • Cognac (brandy) by HS-GC-MS.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

<68.0 °F - closed cup

Flash Point(C)

< 20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization and validation of headspace sorptive extraction for the analysis of volatile compounds in wine vinegars
Callejon RM, et al.
Journal of Chromatography A, 1204(1), 93-103 (2008)
Volatile and sensory profile of organic red wines produced by different selected autochthonous and commercial Saccharomyces cerevisiae strains
Callejon RM, et al.
Analytica Chimica Acta, 660(1-2), 68-75 (2010)
Differences in concentration of principal volatile constituents in traditional Greek distillates
Apostolopoulou AA, et al.
Food Control, 16(2), 157-164 (2005)
Synthesis of diethylacetal: thermodynamic and kinetic studies
Silva VMTM and Rodrigues AE
Chemical Engineering Science, 56(4), 1255-1263 (2001)
Fast determination of principal volatile compounds in distilled spirits.
Lopez-Vazquez C, et al.
Food Control, 21(11), 1436-1441 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service