Skip to Content
Merck
All Photos(1)

Documents

55608

Supelco

Vitexin 2-O-rhamnoside

analytical standard

Synonym(s):

2-O-Rhamnosylvitexin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O14
CAS Number:
Molecular Weight:
578.52
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (TLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2c3c(O)cc(O)c4C(=O)C=C(Oc34)c5ccc(O)cc5)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H30O14/c1-9-19(33)21(35)23(37)27(38-9)41-26-22(36)20(34)16(8-28)40-25(26)18-13(31)6-12(30)17-14(32)7-15(39-24(17)18)10-2-4-11(29)5-3-10/h2-7,9,16,19-23,25-31,33-37H,8H2,1H3/t9-,16+,19-,20+,21+,22-,23+,25-,26+,27-/m0/s1

InChI key

LYGPBZVKGHHTIE-HUBYJIGHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vitexin 2-O-rhamnoside is an active component of hawthorn, a member of the Rosaceae family, exhibits a broad-spectrum of pharmacological properties such as anti-inflammatory, sedative, anti-arrhythmia, effectively inhibiting lipid oxidation, decreasing blood pressure and serum levels of cholesterol, improving digestion and relieving food stasis, etc.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Vitexin 2-O-rhamnoside may be used as an analytical reference standard for the quantification of the analyte in hawthorn leaves, fruits and plasma samples using various chromatography techniques.

Other Notes

This compound is commonly found in plants of the genus: crataegus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation and application by HPLC for simultaneous determination of vitexin-2 ?-O-glucoside, vitexin-2 ?-O-rhamnoside, rutin, vitexin, and hyperoside
Wang H-C, et al.
Journal of Pharmaceutical Analysis, 1(4), 291-296 (2011)
Determination of vitexin-2 ?-O-rhamnoside in rat plasma by ultra-performance liquid chromatography electrospray ionization tandem mass spectrometry and its application to pharmacokinetic study
Ying X, et al.
Talanta, 72(4), 1500-1506 (2007)
Anne Pick et al.
Bioorganic & medicinal chemistry, 19(6), 2090-2102 (2011-03-01)
Flavonoids are an interesting group of natural products ubiquitously present in human diet. Their consumption has been associated with various and differing beneficial health effects. However, several flavonoids have been reported to inhibit the breast cancer resistance protein (BCRP) encoded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service