Skip to Content
Merck
All Photos(1)

Key Documents

55605

Sigma-Aldrich

4-Hydroxy-3-methoxyphenylglycol hemipiperazinium salt

≥98.0% (HPLC)

Synonym(s):

3-Methoxy-4-hydroxy-phenylglycol piperazine, MHPG piperazine, MOPEG, MOPEG piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O4 · 0.5C4H10N2
CAS Number:
Molecular Weight:
227.26
Beilstein:
7360817
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥98.0% (HPLC)

form

solid

mp

115-119 °C

storage temp.

2-8°C

SMILES string

C1CNCCN1.COc2cc(ccc2O)C(O)CO.COc3cc(ccc3O)C(O)CO

InChI

1S/2C9H12O4.C4H10N2/c2*1-13-9-4-6(8(12)5-10)2-3-7(9)11;1-2-6-4-3-5-1/h2*2-4,8,10-12H,5H2,1H3;5-6H,1-4H2

InChI key

KCECBJRHWSOTMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Synthetic norepinephrine metabolite

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T K Keeton et al.
Science (New York, N.Y.), 211(4482), 586-588 (1981-02-06)
Antibodies that specifically bind the norepinephrine metabolite 3-methoxy-4-hydroxyphenylethyleneglycol (MOPEG) were produced in rabbits after injection of a derivative of MOPEG conjugated with bovine thyroglobulin. A sensitive radioimmunoassay was devised with this antiserum, in which as little as 0.5 nanogram of
J.W. Haas, ed.
MHPG, Basic Mechanisms and Psychopathology (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service