Skip to Content
Merck
All Photos(1)

Key Documents

55397

Supelco

(R)-3-Hydroxybutyric acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O3
CAS Number:
Molecular Weight:
104.10
Beilstein:
1720568
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

mp

49-50 °C (lit.)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H](O)CC(O)=O

InChI

1S/C4H8O3/c1-3(5)2-4(6)7/h3,5H,2H2,1H3,(H,6,7)/t3-/m1/s1

InChI key

WHBMMWSBFZVSSR-GSVOUGTGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-3-Hydroxybutyric acid, a monomer of the polymer polyhydroxybutyrate (PHB), is widely used as a nutrition source and a precursor for vitamins, antibiotics and pheromones. Its potential as a protective agent against neuronal death is investigated.

Application

2(R)-3-Hydroxybutyric acid may be used as an analytical reference standard for the quantification of the analyte in urine samples using achiral dual-capillary column gas chromatography. It may also be used as a standard in the production of enantiopure (R)-3-HB using non-conventional yeast A. adeninivorans as a production host and glucose as a carbon source. The purity is determined using gas chromatography-mass spectrometric analysis (GC-MS).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of (R)-3-hydroxybutyric acid by Arxula adeninivorans
Biernack M, et al.
AMB Express, 7(1), 4-4 (2017)
K R Ki et al.
Journal of chromatography. A, 891(2), 257-266 (2000-10-24)
The simultaneous enantiomeric separation of 30 racemic acids including 24 hydroxy acids in a single analysis is described for the determination of their absolute configurations. It involves the conversion of each enantiomer into diastereomeric O-trifluoroacetylated (-)-menthyl ester for the direct
Severe child form of primary hyperoxaluria type 2-a case report revealing consequence of GRHPR deficiency on metabolism
Konkolova J, et al.
BMC Medical Genetics, 18(1), 59-59 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service