Skip to Content
Merck
All Photos(1)

Documents

55137

Supelco

δ-Damascone

analytical standard

Synonym(s):

1-(2,6,6-Trimethyl-3-cyclohexen-1-yl)-2-buten-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.490-1.500

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC1C(C(C=CC)=O)C(C)(C)CC=C1

InChI

1S/C13H20O/c1-5-7-11(14)12-10(2)8-6-9-13(12,3)4/h5-8,10,12H,9H2,1-4H3

InChI key

XEJGJTYRUWUFFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

δ-Damascone is grouped under the class of volatile compounds. It is used as a flavor and fragrance ingredient in various commercial products.

Application

δ-Damascone may be used as an analytical reference standard for the determination of the analyte in perfume and essential oils by low?temperature plasma ionization mass spectrometry.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low-temperature plasma ionization-mass spectrometry for the analysis of compounds in organic aerosol particles
Spencer SE, et al.
Analytical Chemistry, 87(4), 2249-2254 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service