Skip to Content
Merck
All Photos(1)

Key Documents

53963

Supelco

Theaflavin monogallate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H28O16
CAS Number:
Molecular Weight:
716.60
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

description

Mixture of Theaflavin 3-gallate and Theaflavin 3′-gallate

Assay

≥80% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

InChI

1S/C36H28O16/c37-14-5-20(39)18-10-25(44)33(50-28(18)7-14)12-1-16-17(34-26(45)11-19-21(40)6-15(38)8-29(19)51-34)9-27(46)35(30(16)32(48)24(43)2-12)52-36(49)13-3-22(41)31(47)23(42)4-13/h1-9,25-26,33-34,37-42,44-47H,10-11H2,(H,43,48)/t25-,26?,33-,34?/m1/s1

InChI key

CKDOBTPKUHSESZ-MALJIYBWSA-N

General description

Theaflavin monogallate is a polyphenol,[1] and a naturally available antioxidant belonging to the group of theaflavins.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Theaflavin monogallate may be used as a reference standard in the determination of theaflavin monogallate in tea using reversed-phase high-performance liquid chromatography (RP-HPLC) and capillary electrophoresis (CE).[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario A Vermeer et al.
Journal of agricultural and food chemistry, 56(24), 12031-12036 (2008-12-04)
Tea is one of the most widely consumed beverages in the world and may be associated with reduced heart disease rates. Theaflavins, which are formed in the production of black tea, have been suggested being responsible for the blood-cholesterol-lowering (BCL)
G Y Yang et al.
Carcinogenesis, 21(11), 2035-2039 (2000-11-04)
The biological activities of theaflavin (TF), theaflavin gallate (TFG) and theaflavin digallate (TFdiG) from black tea and (-)-epigallocatechin 3-gallate (EGCG) and (-)-epigallocatechin (EGC) from green tea were investigated using SV40-immortalized (33BES) and Ha-ras gene transformed (21BES) human bronchial epithelial cell
Jiebo Lu et al.
European journal of pharmacology, 601(1-3), 66-72 (2008-11-11)
We have previously reported that black tea polyphenol theaflavin monogallate (TF-2) suppressed COX-2 and induced apoptosis in human colon cancer cells [Lu, J.B., Ho, C.-T., Ghai, G., Chen, K.Y., 2000. Differential effects of theaflavin monogallates on cell growth, apoptosis and
Marilisa Leone et al.
Cancer research, 63(23), 8118-8121 (2003-12-18)
Epidemiological data and in vitro studies on cancer chemoprevention by tea polyphenols have gained attention recently from the scientific community, nutritionists, the pharmaceutical industry, and the public. Despite the several efforts made recently to elucidate the molecular basis for the
Yalun Su et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 27(10), 732-733 (2005-04-27)
Qimen black tea has strong inhibitory activity of canola oil oxidation. Four antioxidant compounds, theaflavin (TF1), theaflavin-3-gallate (TF2A), theaflavin-3'-gallate (TF2B) and theaflavin digallate (TF3), were isolated from acetic acetate extract of black tea by silica gel and sephadex LH-20 column

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service