Skip to Content
Merck
All Photos(1)

Key Documents

53439

Supelco

Glutaconic acid

analytical standard

Synonym(s):

2-Pentenedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH=CHCOOH
CAS Number:
Molecular Weight:
130.10
Beilstein:
1759560
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥96.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

SMILES string

OC(=O)C\C=C\C(O)=O

InChI

1S/C5H6O4/c6-4(7)2-1-3-5(8)9/h1-2H,3H2,(H,6,7)(H,8,9)/b2-1+

InChI key

XVOUMQNXTGKGMA-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glutaconic acid is a short chain carboxylic acid which accumulates in the physiological body fluids in patients with deficiency of glutaryl-CoA dehydrogenase leading to glutaric aciduria type 1 disorder (GA1). It finds applications as a diagnostic biomarker since it is found in increased concentrations in the urine of patients with GA1. It can be detected by gas chromatography-mass spectrometry in biological fluids including urine, cerebrospinal fluid, and serum.

Application

Glutaconic acid may be used as a standard to investigate its role in the induction of the mitochondrial permeability transition (MPT) in the isolated rat liver mitochondria in vitro.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two uneventful pregnancies in a woman with Glutaric aciduria type 1
Stepien MK, et al.
JIMD Reports, 41(4), 29-36 (2018)
Induction of the mitochondrial permeability transition in vitro by short-chain carboxylic acids
Palmeira MC, et al.
Biochemical and Biophysical Research Communications, 272, 431-435 (2018)
Prevention of cerebral palsy in glutaric aciduria type 1 by dietary management
Monavari AA and Naugten RE
Archives of Disease in Childhood, 82(1), 67-70 (2000)
Diagnosis and management of glutaric aciduria type I
Baric I, et al.
Journal of inherited metabolic disease, 21(4), 326-340 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service