Skip to Content
Merck
All Photos(1)

Key Documents

52302

Sigma-Aldrich

1-O-Palmityl-rac-glycerol

≥99.0% (GC)

Synonym(s):

rac-1-Hexadecyl-glycerol, rac-Glycerol-1-hexadecyl ether, DL-α-Hexadecylglycerol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2O(CH2)15CH3
CAS Number:
Molecular Weight:
316.52
Beilstein:
1724514
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥99.0% (GC)

mp

60-65 °C

SMILES string

CCCCCCCCCCCCCCCCOCC(O)CO

InChI

1S/C19H40O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-22-18-19(21)17-20/h19-21H,2-18H2,1H3

InChI key

OOWQBDFWEXAXPB-UHFFFAOYSA-N

Gene Information

rat ... Cnr1(25248)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Maulik et al.
Journal of cardiovascular pharmacology, 24(3), 486-492 (1994-09-01)
A recent study demonstrated biochemical and structural alterations of peroxisomes in rat kidney after ischemia/reperfusion. We examined whether peroxisomes play any role in the pathophysiology of myocardial ischemia/reperfusion injury. Isolated perfused rat heart was made ischemic for 30 min by
L E Bertello et al.
Lipids, 32(8), 907-911 (1997-08-01)
A simple method is presented to esterify 1-O-hexadecyl-rac-glycerol using lipases in different organic solvents. The following fatty acids were used: C14:0, C16:0, C18:0, C18:1, and C18:2. Monoesterification was achieved by using a limiting amount of fatty acid. Both the 1-O-hexadecyl-3-O-acylglycerol
S Umeki et al.
Biochimica et biophysica acta, 793(1), 123-128 (1984-03-27)
Exposure of Tetrahymena pyriformis NT-1 to chimyl alcohol (1-O-hexadecyl glycerol) produced a reproducible enhancement in unsaturated fatty acids and a great decrease in order parameter (S), which result from the 2-fold increases of stearoyl-CoA and oleoyl-CoA desaturase activities in microsomes.
B Rüstow et al.
The Biochemical journal, 302 ( Pt 3), 665-668 (1994-09-15)
Alveolar surfactant (exposed to air and therefore a prime target of air oxidants) is supplied with antioxidants during its intracellular formation on type-II pneumocytes [Rüstow, Haupt, Stevens and Kunze (1993) Am. J. Physiol. 265, L133-L139]. Plasmalogens can protect animal cells
A Rait et al.
Bioconjugate chemistry, 11(2), 153-160 (2000-03-22)
Activation of the ras oncogene has been implicated in many types of human tumors. It has been shown that downmodulation of ras expression can lead to the reversion of the transformed phenotype of these tumor cells. Antisense oligodeoxyribonucleotides (ODNs) can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service