Skip to Content
Merck
All Photos(2)

Key Documents

52256

Supelco

(±)-Diclofop

PESTANAL®, analytical standard

Synonym(s):

(RS)-2-[4-(2,4-Dichlorophenoxy)phenoxy]propionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$141.00

$141.00


Estimated to ship onApril 25, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$141.00

About This Item

Empirical Formula (Hill Notation):
C15H12Cl2O4
CAS Number:
Molecular Weight:
327.16
Beilstein:
2338390
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$141.00


Estimated to ship onApril 25, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

composition

carbon content, 54.0-56.2%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(Oc1ccc(Oc2ccc(Cl)cc2Cl)cc1)C(O)=O

InChI

1S/C15H12Cl2O4/c1-9(15(18)19)20-11-3-5-12(6-4-11)21-14-7-2-10(16)8-13(14)17/h2-9H,1H3,(H,18,19)

InChI key

OOLBCHYXZDXLDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service