Skip to Content
Merck
All Photos(1)

Key Documents

51599

Supelco

Spinosad

reference material

Synonym(s):

Mixture of Spinosyn A and Spinosyn D

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107

grade

reference material

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C42H71NO9/c1-10-27-12-11-13-35(52-38-17-16-34(43(5)6)25(4)49-38)24(3)39(45)33-21-31-29(32(33)22-37(44)51-27)15-14-26-19-28(20-30(26)31)50-36-18-23(2)40(46-7)42(48-9)41(36)47-8/h23-36,38,40-42H,10-22H2,1-9H3

InChI key

RQOIAWYOVOXMST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service