Skip to Content
Merck
All Photos(1)

Documents

50803

Supelco

Hexylamine

analytical standard

Synonym(s):

1-Aminohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5NH2
CAS Number:
Molecular Weight:
101.19
Beilstein:
1731298
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

expl. lim.

2.1-9.3 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.417-1.419
n20/D 1.418 (lit.)

bp

131-132 °C (lit.)

mp

−23 °C (lit.)

density

0.766 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCN

InChI

1S/C6H15N/c1-2-3-4-5-6-7/h2-7H2,1H3

InChI key

BMVXCPBXGZKUPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Z Zhou et al.
Oncogene, 34(14), 1831-1842 (2014-05-20)
Extracellular ATP has been shown to either inhibit or promote cancer growth and migration; however, the mechanism underlying this discrepancy remained elusive. Here we demonstrate the divergent roles of ATP and adenosine released by bone osteocytes on breast cancers. We
Maria Notou et al.
Journal of chromatography. A, 1356, 272-276 (2014-07-06)
A novel zone-fluidics/high pressure liquid chromatographic (ZF-HPLC) method is described for the simultaneous determination of six biogenic monoamines, in the presence of hexylamine as internal standard. Automated on-line derivatization of the analytes with naphthalene-2,3-dicarboxaldehyde/cyanide ions was performed using the ZF
Tomáš Taubner et al.
International journal of biological macromolecules, 72, 11-18 (2014-08-12)
Carboxymethylcellulose (CMC) was selected as substrate for amidation based on previous results described for monocarboxy cellulose (MCC) with the aim to prepare highly substituted products. In comparison with MCC containing uronic carboxyl groups at C-6 position, O-carboxymethyl groups in CMC
Ilaria Naldi et al.
PloS one, 9(5), e98101-e98101 (2014-05-24)
Epigenetic events are critical contributors to the pathogenesis of cancer, and targeting epigenetic mechanisms represents a novel strategy in anticancer therapy. Classic demethylating agents, such as 5-Aza-2'-deoxycytidine (Decitabine), hold the potential for reprograming somatic cancer cells demonstrating high therapeutic efficacy
Tamar L Greaves et al.
Physical chemistry chemical physics : PCCP, 17(4), 2357-2365 (2014-12-06)
A high-throughput approach was developed in order to prepare and dry a series of protic ionic liquids (PILs) from 48 Brønsted acid-base combinations. Many combinations comprised an alkyl carboxylic acid paired with an alkyl amine. Visual screens were developed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service