Skip to Content
Merck
All Photos(1)

Key Documents

50738

Millipore

GN Enrichment Broth

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Gram Negative Broth

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$163.00

$163.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$163.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

$163.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

shelf life

limited shelf life, expiry date on the label

composition

dipotassium hydrogen phosphate, 4 g/L
D(+)-glucose, 1 g/L
D-mannitol, 2 g/L
mixed peptone, 20 g/L
potassium dihydrogen phosphate, 1.5 g/L
sodium chloride, 5 g/L
sodium citrate, 5 g/L
sodium deoxycholate, 0.5 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.0±0.2 (37 °C)

application(s)

microbiology

Application

For the selective cultivation of gram-negative intestinal bacteria in particular shigellae from all types of material, acc. to Hajna (1955).
Recommended by the "Schweizerisches Lebensmittelbuch" 5th ed., chapter 56A.

Other Notes

Comparison of selective media for coliphage assay in sewage effluent and lake water[1]

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Kennedy et al.
Applied and environmental microbiology, 49(1), 33-36 (1985-01-01)
Selective media, including EC medium, gram-negative broth, nutrient broth (with 0.05% sodium deoxycholate), and lactose broth (with 0.05% sodium deoxycholate), as well as nonselective nutrient and lactose broths, were compared for the enumeration of coliphages by the agar layer method

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service