Skip to Content
Merck
All Photos(1)

Documents

49968

Supelco

Luteolin 7-glucoside

analytical standard

Synonym(s):

Luteolin 7-O-β-D-glucoside, 3′,4′,5,7-Tetrahydroxyflavone 7-glucoside, Cynaroside, Glucoluteolin, Glucosylluteolin, Luteoloside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](Oc2cc(O)c3C(=O)C=C(Oc3c2)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O11/c22-7-16-18(27)19(28)20(29)21(32-16)30-9-4-12(25)17-13(26)6-14(31-15(17)5-9)8-1-2-10(23)11(24)3-8/h1-6,16,18-25,27-29H,7H2/t16-,18-,19+,20-,21-/m1/s1

InChI key

PEFNSGRTCBGNAN-QNDFHXLGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Luteolin 7-glucoside is a naturally occurring flavanoid isolated from Reseda luteola L.

Application

Luteolin 7-glucoside may be used as an analytical reference standard for the quantification of the anayte in Olea europaea leaves using high-performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: achillea crataegus humulus hypericum melissa mentha pimpinella salvia thymus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and quantitative HPLC analysis of the main flavonoids present in weld (Reseda luteola L.)
Cristea D, et al.
Dyes and Pigments, 57(3), 267-272 (2003)
Rapid high-performance liquid chromatography analysis for the quantitative determination of oleuropein in Olea europaea leaves
Savournin C, et al.
Journal of Agricultural and Food Chemistry, 49(2), 618-621 (2001)
Xiao Sun et al.
Journal of cellular biochemistry, 112(8), 2019-2029 (2011-03-30)
Flavonoids with potent anti-oxidative effects are the major effective components in traditional herbal medicine used in treating cardiovascular diseases. Cynaroside is a flavonoid compound that exhibits anti-oxidative capabilities. However, little is known about its effect on oxidative injury to cardiac
Meihua Jin et al.
Biological & pharmaceutical bulletin, 32(9), 1500-1503 (2009-09-02)
Previously, we reported that an ethanol extract of Ailanthus altissima has antiinflammatory activity in an ovalbumin (OVA)-sensitized murine asthmatic model. To determine the biological compounds from this plant, luteolin-7-O-glucoside (L7G) was isolated and its antiasthmatic activity was evaluated in an
Yihui Yang et al.
Lipids in health and disease, 11, 38-38 (2012-03-15)
In folklore, some people take the decoction of Calyx seu Fructus Physalis (CSFP) for lowering blood lipids. The present study is designed to evaluate the lipid-lowering activities of CSFP, and search for its pharmacodynamical material. CSFP was extracted by water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service