Skip to Content
Merck
All Photos(1)

Key Documents

49497

Supelco

Dimethyl sulfate

for GC derivatization, LiChropur, ≥99.0% (GC)

Synonym(s):

Sulfuric acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2SO2
CAS Number:
Molecular Weight:
126.13
Beilstein:
635994
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

vapor density

4.3 (vs air)

vapor pressure

0.7 mmHg ( 25 °C)

Assay

≥99.0% (GC)

autoignition temp.

923 °F

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Alkylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.386 (lit.)
n20/D 1.387

bp

188 °C (lit.)

mp

−32 °C (lit.)

density

1.333 g/mL at 25 °C (lit.)

SMILES string

COS(=O)(=O)OC

InChI

1S/C2H6O4S/c1-5-7(3,4)6-2/h1-2H3

InChI key

VAYGXNSJCAHWJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dimethyl sulfate may be used as a derivatization reagent for the esterification reaction in the determination of pharmaceuticals by automated aqueous derivatization solid-phase microextraction (SPME) coupled with gas chromatography/mass spectrometry (GC/MS).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M J Cardador et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(21), 1824-1830 (2010-06-15)
Haloacetic acids (HAAs) are water disinfection byproducts (DBPs) formed by the reaction of chlorine oxidizing compounds with natural organic matter in water containing bromine. HAAs are second to trihalomethanes as the most commonly detected DBPs in surface drinking water and
Siming Huang et al.
Talanta, 136, 198-203 (2015-02-24)
An automated aqueous derivatization solid-phase microextraction (SPME) coupled with a gas chromatography/mass spectrometry (GC/MS) method was developed for simultaneous determination of eight pharmaceuticals in water samples. Dimethyl sulfate and tetrabutylammonium hydrogen sulfate were selected as derivatization and activation reagents for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service