Skip to Content
Merck
All Photos(1)

Key Documents

49322

Sigma-Aldrich

NMP

puriss. p.a., ≥99.0% (GC)

Synonym(s):

1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
Beilstein:
106420
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
technique(s):
GC/GC: suitable
bp:
202 °C (lit.)
81-82 °C/10 mmHg (lit.)
vapor pressure:
0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)

Product Name

1-Methyl-2-pyrrolidinone, puriss. p.a., ≥99.0% (GC)

vapor density

3.4 (vs air)

vapor pressure

0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)

grade

puriss. p.a.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

518 °F

expl. lim.

9.5 %

technique(s)

GC/GC: suitable

impurities

≤0.1% water

refractive index

n20/D 1.47 (lit.)
n20/D 1.470

pH

7.7-8

bp

202 °C (lit.)
81-82 °C/10 mmHg (lit.)

mp

−24 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤1 mg/kg

SMILES string

CN1CCCC1=O

InChI

1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3

InChI key

SECXISVLQFMRJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun Ping Neo et al.
Journal of agricultural and food chemistry, 62(22), 5163-5172 (2014-05-16)
Effects of heat treatment on structure and physicochemical properties of zein (Ze) and gallic acid loaded zein (Ze-GA) electrospun fiber mats were investigated. The electrospun fiber mats displayed different surface and physicochemical properties after being heat-cured at 150 °C for
Yanying Lu et al.
Nanoscale, 7(6), 2770-2776 (2015-01-15)
In this paper, we report on the synthesis of micro-nanostructured CuO/C spheres by aerosol spray pyrolysis and their application as high-performance anodes in sodium-ion batteries. Micro-nanostructured CuO/C spheres with different CuO contents were synthesized through aerosol spray pyrolysis by adjusting
B A Jönsson et al.
Journal of chromatography. B, Biomedical sciences and applications, 694(2), 351-357 (1997-07-04)
A method for simultaneous determination of 5-hydroxy-N-methylpyrrolidone and 2-hydroxy-N-methylsuccinimide in urine is described. These compounds are metabolites of N-methyl-2-pyrrolidone, a powerful and widely used organic solvent. 5-Hydroxy-N-methylpyrrolidone and 2-hydroxy-N-methylsuccinimide were purified from urine by adsorption to a C8 solid-phase extraction
Laura Vogelaar et al.
Small (Weinheim an der Bergstrasse, Germany), 1(6), 645-655 (2006-12-29)
Phase separation micromolding (PSmicroM) is a versatile microfabrication technique that can be used to structure a very broad range of polymers, including block copolymers and biodegradable and conductive polymers without the need for clean-room facilities. By incorporating a subsequent process
Ajit Dhananjay Jagtap et al.
European journal of medicinal chemistry, 85, 268-288 (2014-08-05)
A series of 6-acylureido derivatives containing a 3-(pyrrol-2-ylmethylidene)indolin-2-one scaffold were synthesized as potential dual Aurora B/FLT3 inhibitors by replacing the 6-arylureido moiety in 6-arylureidoindolin-2-one-based multi-kinase inhibitors. (Z)-N-(2-(pyrrolidin-1-yl)ethyl)-5-((6-(3-(2-fluoro-4-methoxybenzoyl)ureido)-2-oxoindolin-3-ylidene)methyl)-2,4-dimethyl-1H-pyrrole-3-carboxamide (54) was identified as a dual Aurora B/FLT3 inhibitor (IC50 = 0.4 nM and 0.5 nM, respectively).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service